Learn More
Cayman Chemical C16 (2'(S)-hydroxy) Ceramide (d18:1/16:0), Purity: ≥95%, CAS: 890041-50-6, 25 mg
SDP
Catalog No. 502964109
Encompass_Preferred
A synthetic ceramide; has no effect on C6 glioma cell viability at concentrations up to 20 µM
Catalog No. 50-296-4109 Supplier Cayman Chemical Supplier No. 2251625 MG
A synthetic ceramide; has no effect on C6 glioma cell viability at concentrations up to 20 µM