Learn More
ATCC C166; Yolk-Sac Endothelial Cells; Mouse (Mus musculus)
Catalog No. 502382006
Encompass
C166 is an endothelial cell line that was isolated from the yolk sac of a 12-day-old, mouse embryo. This cell line was deposited by R Auerbach and can be used in cardiovascular disease and stem cell research.
Catalog No. 50-238-2006 Supplier ATCC Supplier No. CRL2581
C166 is an endothelial cell line that was isolated from the yolk sac of a 12-day-old, mouse embryo. This cell line was deposited by R Auerbach and can be used in cardiovascular disease and stem cell research.