Learn More
Aviva Systems Biology Corp C16ORF44 Antibody - N-terminal region (ARP34616_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34616_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against C16ORF44. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C16ORF44. Alternate names: C16orf44ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55481