Learn More
Aviva Systems Biology Corp C16orf57 Antibody - C-terminal region (ARP62380_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62380_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C16orf57. It was validated on Western Blot. . Alternate names: FLJ13154, HVSL1, PN, C16orf57ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-665