Learn More
Aviva Systems Biology Corp C16orf71 Antibody - middle region (ARP52676_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52676_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C16orf71. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C16orf71. Alternate names: DKFZp686H2240, FLJ43261ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60801