Learn More
Aviva Systems Biology Corp C16orf73 Antibody - middle region (ARP55534_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55534_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C16orf73. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C16orf73. Alternate names: MGC35212, gs129, C16orf73ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-052