Learn More
Aviva Systems Biology Corp CFAP20 Antibody - middle region (ARP34302_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34302_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C16orf80. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C16orf80. Alternate names: EVORF, GTL3, fSAP23ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55332