Learn More
Cayman Chemical C17 CeramIded18 117 0 10mg
SDP
Catalog No. 502986767
Encompass_Preferred
A synthetic ceramide; contains a non-natural C17:0 fatty acid acylated to sphingosine; has been used as a marker of arylsulfatase A and sphingolipid activator B activity
Catalog No. 50-298-6767 Supplier Cayman Chemical Supplier No. 2253210 MG
A synthetic ceramide; contains a non-natural C17:0 fatty acid acylated to sphingosine; has been used as a marker of arylsulfatase A and sphingolipid activator B activity