Learn More
Cayman Chemical C17 Ceramide (d18:1/17:0), Purity: ≥98%, CAS: 67492-16-4, 5 mg
SDP
Catalog No. 502987735
Encompass_Preferred
A synthetic ceramide; contains a non-natural C17:0 fatty acid acylated to sphingosine; has been used as a marker of arylsulfatase A and sphingolipid activator B activity
Catalog No. 50-298-7735 Supplier Cayman Chemical Supplier No. 225325 MG
A synthetic ceramide; contains a non-natural C17:0 fatty acid acylated to sphingosine; has been used as a marker of arylsulfatase A and sphingolipid activator B activity