Learn More
Cayman Chemical C17 DerythroLthreo Sphngo 10mg
SDP
Catalog No. 502936508
Encompass_Preferred
A synthetic derivative of sphingomyelin; has been used as an internal standard for the quantification of sphingomyelin
Catalog No. 50-293-6508 Supplier Cayman Chemical Supplier No. 2435410 MG
A synthetic derivative of sphingomyelin; has been used as an internal standard for the quantification of sphingomyelin