Learn More
Cayman Chemical C17 Globotriaosylceramide d1
SDP
Catalog No. 503016011
Encompass_Preferred
A sphingolipid; has been used as an internal standard for the quantification of globotriaosylceramide in plasma and urine from patients with Fabry disease by GC-MS
Catalog No. 50-301-6011 Supplier Cayman Chemical Supplier No. 24876500 UG
A sphingolipid; has been used as an internal standard for the quantification of globotriaosylceramide in plasma and urine from patients with Fabry disease by GC-MS