Learn More
Medchemexpress LLC C17H11Br2NO2S2 | 893449-38-2 | 99.1% | 485.21 | 10 MG
SDP
Catalog No. 5000438636
Encompass_Preferred
PRL-3 Inhibitor I is a potent PRL-3 inhibitor with an IC50 of 0.9 μM. It demonstrates reduced invasion in cell-based assays.
  • Potent PRL-3 inhibitor with an IC50 of 0.9 μM.
  • Reduces invasion in cell-based assay.
  • For research use only.
Catalog No. 50-004-38636 Supplier Medchemexpress LLC Supplier No. HY11247610MG
PRL-3 Inhibitor I is a potent PRL-3 inhibitor with an IC50 of 0.9 μM. It demonstrates reduced invasion in cell-based assays.
  • Potent PRL-3 inhibitor with an IC50 of 0.9 μM.
  • Reduces invasion in cell-based assay.
  • For research use only.