Learn More
Aviva Systems Biology Corp C17orf39 Antibody - C-terminal region (ARP53761_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53761_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C17orf39. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human C17orf39. Alternate names: MGC3048, C17orf39ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61119