Learn More
Aviva Systems Biology Corp C17orf48 Antibody - middle region (ARP57377_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57377_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C17orf48. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C17orf48. Alternate names: MDS006, NBLA03831, C17orf48ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54009