Learn More
Aviva Systems Biology Corp C17orf64 Antibody - middle region (ARP53402_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53402_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C17orf64. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C17orf64. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Yeast
Catalog No. 50-014-61017