Learn More
Aviva Systems Biology Corp HEATR9 Antibody - C-terminal region (ARP63149_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63149_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C17orf66. It was validated on Western Blot. . Alternate names: FLJ32830ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Rat
Catalog No. 50-287-895