Learn More
Aviva Systems Biology Corp SPEM2 Antibody - middle region (ARP53323_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53323_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C17orf74. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C17orf74. Alternate names: -ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-60999