Learn More
Aviva Systems Biology Corp C17orf75 Antibody - N-terminal region (ARP57623_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57623_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C17orf75. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C17orf75. Alternate names: NJMU-R1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-62913