Learn More
Aviva Systems Biology Corp C17orf80 Antibody - N-terminal region (ARP47381_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47381_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C17orf80. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C17orf80. Alternate names: FLJ20721, HLC-8, MIG3, SPEP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59531