Learn More
Avanti Polar Lipids C18:0-d7 GD3 Ceramide (d18:1/18:0-d7), synthetic, 100ug
Catalog No. NC3669224
Encompass
GD3 ceramide is the main ganglioside within human breast milk. GD3 Ceramides are critical for apoptosis and present in high quantities in various tumor species. This GD2 Ceramide is the deuterated version, which will enable the easier study of structure, dynamics, and function of cell membranes.
Catalog No. NC3669224 Supplier Avanti Polar Lipids Supplier No. 860123W100UG
GD3 ceramide is the main ganglioside within human breast milk. GD3 Ceramides are critical for apoptosis and present in high quantities in various tumor species. This GD2 Ceramide is the deuterated version, which will enable the easier study of structure, dynamics, and function of cell membranes.