Learn More
Avanti Polar Lipids C18:0-d7 GT3 Ceramide (d18:1/18:0-d7), synthetic, 100ug
Catalog No. NC3669223
Encompass
GT3 is a ceramide that is predominantly found in the retina and may play a role in retina tissue structure and function. GT3 ceramides are precursors to more complex gangliosides, and modified GT3 ceramides have been linked to tumors. Lower levels of GT3 ceramides are seen in patients with Alzheimer’s disease. This GT3 ganglioside is the deuterated version, which will enable the easier study of structure, dynamics, and function of cell membranes.
Catalog No. NC3669223 Supplier Avanti Polar Lipids Supplier No. 860126W100UG
GT3 is a ceramide that is predominantly found in the retina and may play a role in retina tissue structure and function. GT3 ceramides are precursors to more complex gangliosides, and modified GT3 ceramides have been linked to tumors. Lower levels of GT3 ceramides are seen in patients with Alzheimer’s disease. This GT3 ganglioside is the deuterated version, which will enable the easier study of structure, dynamics, and function of cell membranes.