Learn More
Medchemexpress LLC C18:1-Ceramide | 5966-28-9 | 99.9% | C36H69NO3 | 50 MG
SDP
Catalog No. 5000357042
Encompass_Preferred
C18:1-Ceramide is a ceramide subspecies primarily used for research and analytical applications. It serves as an analytical standard and can be utilized in studies related to type 2 diabetes.
  • Ceramide subspecies
  • Intended for research and analytical applications
  • Can be used in the study of type 2 diabetes
Catalog No. 50-003-57042 Supplier Medchemexpress LLC Supplier No. HY14157850MG
C18:1-Ceramide is a ceramide subspecies primarily used for research and analytical applications. It serves as an analytical standard and can be utilized in studies related to type 2 diabetes.
  • Ceramide subspecies
  • Intended for research and analytical applications
  • Can be used in the study of type 2 diabetes