Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avanti Polar Lipids C18:1 Glucosyl(ß) Ceramide, 5 mg, Powder, C42H79NO8, Glycosphingolipid, Ceramide, 726.079 g/mol, β-1,1' Glucosyl Linkage, 9Z-octadecenoyl (Oleoyl)

Catalog No. NC1439548
Encompass

C18:1 Glucosyl(ß) Ceramide (d18:1/18:1(9Z)) is a high-purity glycosphingolipid from Avanti Polar Lipids. This ceramide consists of an oleoyl fatty acid chain and a D-erythro-sphingosine base, and is available in a 5 mg quantity. The empirical formula is C42H79NO8, with a molecular weight of 726.079, making it valuable for lipid biology research.

  • High-purity glycosphingolipid
  • Available in 5 mg quantity
  • Empirical formula: C42H79NO8
  • Molecular weight: 726.079
  • Lipid class: Glycosphingolipid, Ceramide
  • Fatty acid: 9Z-octadecenoyl (Oleoyl)
  • Sphingoid base: D-erythro-sphingosine (d18:1)
  • Physical form: Powder

Catalog No. NC1439548 Supplier Avanti Polar Lipids Supplier No. 860548P5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C18:1 Glucosyl(ß) Ceramide (d18:1/18:1(9Z)) is a high-purity glycosphingolipid from Avanti Polar Lipids. This ceramide consists of an oleoyl fatty acid chain and a D-erythro-sphingosine base, and is available in a 5 mg quantity. The empirical formula is C42H79NO8, with a molecular weight of 726.079, making it valuable for lipid biology research.

  • High-purity glycosphingolipid
  • Available in 5 mg quantity
  • Empirical formula: C42H79NO8
  • Molecular weight: 726.079
  • Lipid class: Glycosphingolipid, Ceramide
  • Fatty acid: 9Z-octadecenoyl (Oleoyl)
  • Sphingoid base: D-erythro-sphingosine (d18:1)
  • Physical form: Powder

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.