Learn More
Sorbent Technologies Inc C18 Silica HPTLC Plates, 100% silanization, glass backed, 200um, 10x10cm, 25pk
SDP
Catalog No. 501138598
Encompass_Preferred
This high performance thin layer chromatography (HPTLC) plate comprises a C18 Silica layer. The Nano-Silica 60 has a surface area of 500 m2/g, mean pore size of 60A, specific pore volume of 0.75 ml/g, particle size of 2-10um, and pH stability of 2-10. The surface has been 100% treated with an octadecyl silane so that it is a reversed phase material. This analytical hard layer is bound to a glass support. It has a thickness of 200um and dimensions of 10x10cm. There are 25 plates per pack. Applications include: Alkaloids, amino acids, preservatives, optical brighteners, barbiturates, polycyclic aromatic hydrocarbons (PAH), drugs, peptides, flavonoids, phenols, indole derivatives, steroids
Catalog No. 50-113-8598 Supplier Sorbent Technologies Inc Supplier No. 2514056
This high performance thin layer chromatography (HPTLC) plate comprises a C18 Silica layer. The Nano-Silica 60 has a surface area of 500 m2/g, mean pore size of 60A, specific pore volume of 0.75 ml/g, particle size of 2-10um, and pH stability of 2-10. The surface has been 100% treated with an octadecyl silane so that it is a reversed phase material. This analytical hard layer is bound to a glass support. It has a thickness of 200um and dimensions of 10x10cm. There are 25 plates per pack. Applications include: Alkaloids, amino acids, preservatives, optical brighteners, barbiturates, polycyclic aromatic hydrocarbons (PAH), drugs, peptides, flavonoids, phenols, indole derivatives, steroids