Learn More
Medchemexpress LLC C18-ceramide-13C2,d2 | 569.95 g·mol⁻1 | C3413C2H69D2NO3 | 1 MG
SDP
Catalog No. 5000284728
Encompass_Preferred
C18-ceramide-13C2,d2 is a stable isotope-labeled C18 ceramide intended for use as an internal standard in lipidomics and mass spectrometry. The dual 13C and deuterium labels provide a defined mass shift to support accurate quantitation and metabolic tracing in analytical workflows.
  • Stable isotope labeled with 13C2 and d2 for clear mass-shifted detection.
  • Suitable as an internal standard for quantitative mass spectrometry workflows.
  • Useful for metabolic tracing and method development in lipidomics.
  • Provided as a small, research-grade quantity for analytical applications.
Catalog No. 50-002-84728 Supplier Medchemexpress LLC Supplier No. HYW6541051MG
C18-ceramide-13C2,d2 is a stable isotope-labeled C18 ceramide intended for use as an internal standard in lipidomics and mass spectrometry. The dual 13C and deuterium labels provide a defined mass shift to support accurate quantitation and metabolic tracing in analytical workflows.
  • Stable isotope labeled with 13C2 and d2 for clear mass-shifted detection.
  • Suitable as an internal standard for quantitative mass spectrometry workflows.
  • Useful for metabolic tracing and method development in lipidomics.
  • Provided as a small, research-grade quantity for analytical applications.