Learn More
Medchemexpress LLC C18-ceramide | 2304-81-6 | 100.0% | 565.95 | C36H71NO3 | 100 MG
SDP
Catalog No. 5000214255
Encompass_Preferred
C18-Ceramide is a research-grade sphingolipid that modulates cellular stress and signaling pathways. It crosses the blood-brain barrier and is commonly used in neuroscience and oncology studies to investigate ER stress, PI3K/AKT signaling, autophagy, and apoptosis.
  • High purity suitable for analytical and biological studies.
  • White to off-white solid; stable under recommended storage conditions.
  • Soluble in ethanol and DMF; limited solubility in DMSO.
  • Available in multiple package sizes for flexible experimental needs.
  • Relevant for studies of neuronal injury and tumor biology.
Catalog No. 50-002-14255 Supplier Medchemexpress LLC Supplier No. HY100355100MG
Add to Cart
Add to Cart
C18-Ceramide is a research-grade sphingolipid that modulates cellular stress and signaling pathways. It crosses the blood-brain barrier and is commonly used in neuroscience and oncology studies to investigate ER stress, PI3K/AKT signaling, autophagy, and apoptosis.
  • High purity suitable for analytical and biological studies.
  • White to off-white solid; stable under recommended storage conditions.
  • Soluble in ethanol and DMF; limited solubility in DMSO.
  • Available in multiple package sizes for flexible experimental needs.
  • Relevant for studies of neuronal injury and tumor biology.