Learn More
Medchemexpress LLC C18-ceramide | 2304-81-6 | 100.0% | 565.95 g/mol | C36H71NO3 | 25 MG
SDP
Catalog No. 5000214256
Encompass_Preferred
C18-ceramide is a saturated sphingolipid research reagent used to probe cellular signaling processes such as apoptosis, autophagy, endoplasmic reticulum stress, and PI3K/AKT pathway modulation. It is supplied as a high-purity powder for in vitro studies and should be stored under recommended conditions to preserve stability.
  • High purity suitable for biochemical and cell-based assays
  • Relevant for neuroscience and oncology research applications
  • Modulates apoptosis, autophagy, and endoplasmic reticulum stress pathways
  • Available in small, laboratory-friendly package sizes
  • Stable when stored under recommended conditions for reproducible results
Catalog No. 50-002-14256 Supplier Medchemexpress LLC Supplier No. HY10035525MG
C18-ceramide is a saturated sphingolipid research reagent used to probe cellular signaling processes such as apoptosis, autophagy, endoplasmic reticulum stress, and PI3K/AKT pathway modulation. It is supplied as a high-purity powder for in vitro studies and should be stored under recommended conditions to preserve stability.
  • High purity suitable for biochemical and cell-based assays
  • Relevant for neuroscience and oncology research applications
  • Modulates apoptosis, autophagy, and endoplasmic reticulum stress pathways
  • Available in small, laboratory-friendly package sizes
  • Stable when stored under recommended conditions for reproducible results