Learn More
Medchemexpress LLC C18-ceramide | 2304-81-6 | 100.0% | 565.95 g/mol | C36H71NO3 | 5 MG
SDP
Catalog No. 5000212700
Encompass_Preferred
C18-ceramide is a naturally occurring sphingolipid used as a research reagent to investigate cell signaling, apoptosis, autophagy, and neuronal injury mechanisms. It is reported to cross the blood-brain barrier and to affect endoplasmic reticulum stress pathways, PI3K/AKT signaling, and membrane trafficking processes.
  • Useful for studies of apoptosis and autophagy
  • Crosses the blood-brain barrier, enabling neuroscience applications
  • Modulates endoplasmic reticulum stress and PI3K/AKT signaling
  • High purity suitable for analytical and biological assays
  • Multiple pack sizes available to match experimental scale
  • Soluble in ethanol and DMF with sonication for in vitro use
Catalog No. 50-002-12700 Supplier Medchemexpress LLC Supplier No. HY1003555MG
C18-ceramide is a naturally occurring sphingolipid used as a research reagent to investigate cell signaling, apoptosis, autophagy, and neuronal injury mechanisms. It is reported to cross the blood-brain barrier and to affect endoplasmic reticulum stress pathways, PI3K/AKT signaling, and membrane trafficking processes.
  • Useful for studies of apoptosis and autophagy
  • Crosses the blood-brain barrier, enabling neuroscience applications
  • Modulates endoplasmic reticulum stress and PI3K/AKT signaling
  • High purity suitable for analytical and biological assays
  • Multiple pack sizes available to match experimental scale
  • Soluble in ethanol and DMF with sonication for in vitro use