Learn More
Medchemexpress LLC C18-ceramide | 2304-81-6 | MFCD00056222 | 100.0% | 565.95 g/mol | C36H71NO3 | 10 MG
SDP
Catalog No. 5000214573
Encompass_Preferred
C18-ceramide is a long-chain sphingolipid used as a biochemical reagent and analytical standard in research on sphingolipid signaling, apoptosis, and membrane biology. Supplied in milligram-scale vials, it is suitable for preparing concentrated stock solutions for cell-based assays, biochemical experiments, and mass spectrometry analysis.
  • High purity analytical standard suitable for research.
  • Available in milligram-scale vials for accurate dosing.
  • Soluble in common organic solvents for stock preparation.
  • Useful in studies of apoptosis, cell signaling, and membrane dynamics.
  • Provided with supporting documentation including a product data sheet and safety information.
Catalog No. 50-002-14573 Supplier Medchemexpress LLC Supplier No. HY10035510MG
C18-ceramide is a long-chain sphingolipid used as a biochemical reagent and analytical standard in research on sphingolipid signaling, apoptosis, and membrane biology. Supplied in milligram-scale vials, it is suitable for preparing concentrated stock solutions for cell-based assays, biochemical experiments, and mass spectrometry analysis.
  • High purity analytical standard suitable for research.
  • Available in milligram-scale vials for accurate dosing.
  • Soluble in common organic solvents for stock preparation.
  • Useful in studies of apoptosis, cell signaling, and membrane dynamics.
  • Provided with supporting documentation including a product data sheet and safety information.