Learn More
Medchemexpress LLC C18-ceramide | 2304-81-6 | MFCD00056222 | 100.0% | 565.95 g/mol | C36H71NO3 | 50 MG
SDP
Catalog No. 5000214254
Encompass_Preferred
C18-ceramide is a naturally occurring bioactive sphingolipid used in biomedical research to investigate cell signaling, apoptosis, autophagy, and neuronal injury mechanisms. It modulates endoplasmic reticulum stress and PI3K/AKT pathways, can influence exocytosis and lipid-raft functions, and is commonly applied in oncology and neuroscience studies. Supplied for research use only.
  • Bioactive sphingolipid involved in apoptosis and autophagy
  • Modulates endoplasmic reticulum stress and PI3K/AKT signaling pathways
  • Capable of crossing the blood-brain barrier
  • High purity (99.95%) suitable for research applications
  • Available in multiple package sizes, including 50 mg
  • Supplied for research use only; not for human use
Catalog No. 50-002-14254 Supplier Medchemexpress LLC Supplier No. HY10035550MG
C18-ceramide is a naturally occurring bioactive sphingolipid used in biomedical research to investigate cell signaling, apoptosis, autophagy, and neuronal injury mechanisms. It modulates endoplasmic reticulum stress and PI3K/AKT pathways, can influence exocytosis and lipid-raft functions, and is commonly applied in oncology and neuroscience studies. Supplied for research use only.
  • Bioactive sphingolipid involved in apoptosis and autophagy
  • Modulates endoplasmic reticulum stress and PI3K/AKT signaling pathways
  • Capable of crossing the blood-brain barrier
  • High purity (99.95%) suitable for research applications
  • Available in multiple package sizes, including 50 mg
  • Supplied for research use only; not for human use