Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C18-ceramide-d7 | 1840942-14-4 | 99.0% | 573.00 | C36H64D7NO3 | 500 UG
SDP

Catalog No. 5000214253
Encompass_Preferred

C18-ceramide-d7 is a deuterium-labeled analogue of C18-ceramide used as a tracer and internal standard in lipid analysis and mass spectrometry. Supplied as a solution at 1.75 mM in methanol:chloroform (10:90), the material is intended for research use only and should be stored at -20°C.

  • Isotopically labeled internal standard (d7) for quantitative lipidomics
  • High purity: 99.0%
  • Molecular weight: 573.00
  • Chemical formula: C36H64D7NO3
  • Pack size: 500 μg, 1.75 mM in 500 μL
  • Storage: store at -20°C

Catalog No. 50-002-14253 Supplier Medchemexpress LLC Supplier No. HY100355S1500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C18-ceramide-d7 is a deuterium-labeled analogue of C18-ceramide used as a tracer and internal standard in lipid analysis and mass spectrometry. Supplied as a solution at 1.75 mM in methanol:chloroform (10:90), the material is intended for research use only and should be stored at -20°C.

  • Isotopically labeled internal standard (d7) for quantitative lipidomics
  • High purity: 99.0%
  • Molecular weight: 573.00
  • Chemical formula: C36H64D7NO3
  • Pack size: 500 μg, 1.75 mM in 500 μL
  • Storage: store at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.