Learn More
Cayman Chemical C18 CeramIde-d7d18 1-d718250ug
SDP
Catalog No. 502993928
Encompass_Preferred
An internal standard for the quantification of C18 ceramide by GC- or LC-MS
Catalog No. 50-299-3928 Supplier Cayman Chemical Supplier No. 22788250 UG
An internal standard for the quantification of C18 ceramide by GC- or LC-MS