Learn More
Cayman Chemical C18 dihydro Ceramide-d3 (d18:0/18:0-d3), Purity: ≥99% deuterated forms (d1-d3); ≤1% d0, CAS: 2699606-99-8, 1 mg
SDP
Catalog No. 502936514
Encompass_Preferred
An internal standard for the quantification of C18 dihydro ceramide by GC- or LC-MS
Catalog No. 50-293-6514 Supplier Cayman Chemical Supplier No. 244281 MG
An internal standard for the quantification of C18 dihydro ceramide by GC- or LC-MS