Learn More
Cayman Chemical C18 dIhydro CRamIDd18 01 10mg
SDP
Catalog No. 503015840
Encompass_Preferred
A bioactive sphingolipid; precursor in the de novo synthesis of C18 ceramide; leads to increased triacylglycerol storage and autophagosome accumulation as well as upregulation of the fibrosis markers α-SMA and FGF2 in Chang and LX-2 liver cells; elevated in the plasma of pre-diabetics and the skin of patients with lesional atopic dermatitis
Catalog No. 50-301-5840 Supplier Cayman Chemical Supplier No. 2437910 MG
A bioactive sphingolipid; precursor in the de novo synthesis of C18 ceramide; leads to increased triacylglycerol storage and autophagosome accumulation as well as upregulation of the fibrosis markers α-SMA and FGF2 in Chang and LX-2 liver cells; elevated in the plasma of pre-diabetics and the skin of patients with lesional atopic dermatitis