Learn More
Cayman Chemical C18 Fatty Acid Methyl Ester Mixture, 1 mL
SDP
Catalog No. 502988129
Encompass_Preferred
A mixture of C18 FAMEs; intended for use as a reference standard for the quantification of these C18 fatty acids in various sample types by GC-FID or GC-MS
Catalog No. 50-298-8129 Supplier Cayman Chemical Supplier No. 293691 ML
A mixture of C18 FAMEs; intended for use as a reference standard for the quantification of these C18 fatty acids in various sample types by GC-FID or GC-MS