Learn More
Cayman Chemical C18 Globotriaosylceramide d1
SDP
Catalog No. 503015869
Encompass_Preferred
A sphingolipid; inhibits PMA-induced aggregation of human neutrophils (1 μM); acts as a receptor for Shiga toxin in B cell-derived Raji cells and THP-1 monocytes; accumulates in the brain, heart, kidney, liver, lung, and spleen in a mouse model of Fabry disease; accumulates in various cells in patients with Fabry disease
Catalog No. 50-301-5869 Supplier Cayman Chemical Supplier No. 24877500 UG
A sphingolipid; inhibits PMA-induced aggregation of human neutrophils (1 μM); acts as a receptor for Shiga toxin in B cell-derived Raji cells and THP-1 monocytes; accumulates in the brain, heart, kidney, liver, lung, and spleen in a mouse model of Fabry disease; accumulates in various cells in patients with Fabry disease