Learn More
Cayman Chemical C18 GlobotrIaosylcRamIDd3
SDP
Catalog No. 502934472
Encompass_Preferred
An internal standard for the quantification of C18 globotriaosylceramide by GC- or LC-MS
Catalog No. 50-293-4472 Supplier Cayman Chemical Supplier No. 24626500 UG
An internal standard for the quantification of C18 globotriaosylceramide by GC- or LC-MS