Learn More
Medchemexpress LLC C18 glucosyl (β) ceramide (d18:1/18:0) | 95119-86-1 | 99.9% | 728.09 | 25 MG

Supplier: Medchemexpress LLC 25MGHY170035
GlcCer (d18:1/18:0), also known as C18 glucosyl (β) ceramide (d18:1/18:0), is a glycosphingolipid. This compound is promising for research into neurodegenerative diseases such as Parkinson's disease and dementia with Lewy bodies.
- Glycosphingolipid compound.
- Suitable for Parkinson's disease research.
- Suitable for dementia with Lewy bodies research.
- Store powder at -20°C for up to 3 years.
- Store in solvent at -80°C for 6 months or -20°C for 1 month.
- Soluble in DMSO at 2.5 mg/mL (ultrasonic needed).
- Appears as a white to off-white solid.
- Purity by HPLC is 99.9%.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.