Learn More
Medchemexpress LLC C18 glucosyl(β) ceramide (d18:1/18:0) | 95119-86-1 | 99.9% | 728.09 g/mol | C42H81NO8 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17003510MG
Encompass_Preferred
GlcCer (d18:1/18:0) is a C18 glucosylceramide glycosphingolipid used as a biochemical research reagent for studies of glycosphingolipid metabolism and neurodegenerative disorders, including Parkinson disease and dementia with Lewy bodies. The compound is supplied as a solid for dissolution in organic solvent and is characterized by formula C42H81NO8 and molecular weight 728.09 g/mol.
  • Used in studies of glycosphingolipid biology and neurodegeneration.
  • Suitable for biochemical and cellular assays.
  • Soluble in DMSO; ultrasonic treatment may be required for full dissolution.
  • Stable as powder at -20°C for extended storage.
  • High purity appropriate for research applications.
Catalog No. 50-002-94926