Learn More
Cayman Chemical C18 Phytoceramide (t18:0/18:0), Purity: ≥95%, CAS: 34354-88-6, 5 mg
SDP
Catalog No. 502994711
Encompass_Preferred
A bioactive sphingolipid found in S. cerevisiae, wheat grains, and mammalian epidermis; has a role in regulation of apoptosis, cell differentiation, proliferation of smooth muscle cells, and inhibition of the mitochondrial respiratory chain; inhibits expression of the allergic cytokines IL-4, TNF-α, and transcription factors c-Jun and NF-κB in histone-stimulated tissue; is used in cosmetics as a skin protectant
Catalog No. 50-299-4711 Supplier Cayman Chemical Supplier No. 226865 MG
A bioactive sphingolipid found in S. cerevisiae, wheat grains, and mammalian epidermis; has a role in regulation of apoptosis, cell differentiation, proliferation of smooth muscle cells, and inhibition of the mitochondrial respiratory chain; inhibits expression of the allergic cytokines IL-4, TNF-α, and transcription factors c-Jun and NF-κB in histone-stimulated tissue; is used in cosmetics as a skin protectant