Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C18(plasm) LPC (PC(18:1p/0:0)) | 97802-55-6 | MFCD00133396 | >99.0% | 507.68 | C26H54NO6P | 1 MG
SDP

Catalog No. 5000300758
Encompass_Preferred

C18(Plasm) LPC (PC(18:1p/0:0)) is a plasmalogen-class lysophosphatidylcholine offered as a high-purity research reagent for lipidomics, metabolic profiling, and analytical method development. Supplied as a powder with a defined molecular formula and molecular weight, it is intended for use as an endogenous metabolite standard and for biochemical assays.

  • High purity (>99.0%) for reliable analytical results.
  • Molecular weight 507.68 facilitating mass spectrometry identification.
  • Powder form stable at -20°C for long-term storage.
  • Solvent storage options for short-term experimental use (-80°C or -20°C).
  • Suitable for lipidomics, metabolism studies, and method development.

Catalog No. 50-003-00758 Supplier Medchemexpress LLC Supplier No. HYW0208091MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C18(Plasm) LPC (PC(18:1p/0:0)) is a plasmalogen-class lysophosphatidylcholine offered as a high-purity research reagent for lipidomics, metabolic profiling, and analytical method development. Supplied as a powder with a defined molecular formula and molecular weight, it is intended for use as an endogenous metabolite standard and for biochemical assays.

  • High purity (>99.0%) for reliable analytical results.
  • Molecular weight 507.68 facilitating mass spectrometry identification.
  • Powder form stable at -20°C for long-term storage.
  • Solvent storage options for short-term experimental use (-80°C or -20°C).
  • Suitable for lipidomics, metabolism studies, and method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.