Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ C18-Reversed Phase Silica Gel, Fully Endcapped, 15-25 μm

Catalog No. 111003871 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3871 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3871 Supplier MilliporeSigma™ Supelco™ Supplier No. 6075650G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For column chromatography

Reversed-phase silica gels are generally suitable to retain catalysts by hydrophobic interactions.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 15 to 25 μm
Pore Size 100 Å
Grade For column chromatography
Quantity 50 g
Surface Area (Metric) 380 m2/g
MDL Number MFCD00148338
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.