Learn More
MilliporeSigma™ Supelco™ C18-Reversed Phase Silica Gel, Fully Endcapped, 230-400 mesh
Catalog No. 111003039 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 g
Catalog No. Quantity
11-100-3039 250 g
1 options
Catalog No. 11-100-3039 Supplier MilliporeSigma™ Supelco™ Supplier No. 60757250G
Add to Cart
Add to Cart
For column chromatography
Reversed-phase silica gels are generally suitable to retain catalysts by hydrophobic interactions.
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 230/400 Mesh
Pore Size 90 Å
Grade For column chromatography
Quantity 250 g
Surface Area (Metric) 400 m2/g