Learn More
Sorbent Technologies Inc C18 Spherical Silica Gel, Premium Rf, 70A, 40-75um, 100g
SDP
Catalog No. 501138865
Encompass_Preferred
C18 Spherical Silica Gel, Premium Rf has a porosity of 70A, particle distribution of 40-75um, and a quantity of 100g. Spherical Silica Gel, Premium Rf when compared to granular silica gels show an increase loading (up to 30% more), greater reproducibility of process, sharper peaks and cost savings (less solvent consumption and labor), longer bed lifetime (decrease cost of labor and silica consumption), and better chromatographic results. This high-purity spherical silica gel is ideal for flash chromatography, industrial scale LC, and batch adsorption applications. C18 is the most popular and versatile reversed phase, capable of hydrophobic interactions with a wide variety of organic compounds. It is used for separation of nonpolar to moderately polar compounds such as: Fatty Acids, Glycerides, Polycyclic Aromatics, Esters (Phthalates), Fat-Soluble Vitamins, Steroids, Prostagl&ins, PTH amino acids.
Catalog No. 50-113-8865 Supplier Sorbent Technologies Inc Supplier No. 6254601
C18 Spherical Silica Gel, Premium Rf has a porosity of 70A, particle distribution of 40-75um, and a quantity of 100g. Spherical Silica Gel, Premium Rf when compared to granular silica gels show an increase loading (up to 30% more), greater reproducibility of process, sharper peaks and cost savings (less solvent consumption and labor), longer bed lifetime (decrease cost of labor and silica consumption), and better chromatographic results. This high-purity spherical silica gel is ideal for flash chromatography, industrial scale LC, and batch adsorption applications. C18 is the most popular and versatile reversed phase, capable of hydrophobic interactions with a wide variety of organic compounds. It is used for separation of nonpolar to moderately polar compounds such as: Fatty Acids, Glycerides, Polycyclic Aromatics, Esters (Phthalates), Fat-Soluble Vitamins, Steroids, Prostagl&ins, PTH amino acids.