Learn More
Cayman Chemical C18 Sphingomyelin (d18:1/18:0), Purity: ≥98%, CAS: 54336-69-5, 5 mg
SDP
Catalog No. 502932989
Encompass_Preferred
A bioactive sphingolipid; naturally occurring form of sphingomyelin; increased by approximately 35-fold in CCD-112 colon fibroblasts compared with HCT116 human colorectal carcinoma cells; has been used as an internal standard for the quantification of sphingolipid metabolites; a mixture of the D-erythro and L-threo isomers of C18 sphingomyelin
Catalog No. 50-293-2989 Supplier Cayman Chemical Supplier No. 243555 MG
A bioactive sphingolipid; naturally occurring form of sphingomyelin; increased by approximately 35-fold in CCD-112 colon fibroblasts compared with HCT116 human colorectal carcinoma cells; has been used as an internal standard for the quantification of sphingolipid metabolites; a mixture of the D-erythro and L-threo isomers of C18 sphingomyelin