Learn More
Waters Corp VanGuard Cartridge, C18, 90 Å, 2.7 μm, 2.1 mm x 5 mm
Catalog No. NC2455458 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by removing particulate contamination from the mobile phase stream. This cartridge is specifically optimized to protect all 2.1 mm and 3.0 mm I.D. analytical columns that contain 2.7 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized to protect 2.1 mm and 3.0 mm I.D. analytical columns with 2.7 μm sorbent particles.
  • Suitable for physical and chemical protection of analytical columns.
  • Offers cost-effective protection by eliminating particulates and chemical contamination.
Catalog No. NC2455458 Supplier Waters Corp Supplier No. 186007685
VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by removing particulate contamination from the mobile phase stream. This cartridge is specifically optimized to protect all 2.1 mm and 3.0 mm I.D. analytical columns that contain 2.7 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized to protect 2.1 mm and 3.0 mm I.D. analytical columns with 2.7 μm sorbent particles.
  • Suitable for physical and chemical protection of analytical columns.
  • Offers cost-effective protection by eliminating particulates and chemical contamination.