Learn More
Medchemexpress LLC HY-112288 10mg , C188-9 CAS:432001-19-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11228810MG
Encompass_Preferred
HY-112288 10mg C188-9 CAS: 432001-19-9 C188-9 (TTI-101) is a STAT3 inhibitor, with a Kd of 4.7 nM. C188-9 inhibits G-CSF-induced STAT3 activation and STAT3-dependent gene expression. C188-9 induces apoptosis in AML cell lines and primary samples and inhibits colony formation by primary AML blasts[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9631