Learn More
Medchemexpress LLC HY-112288 5mg Medchemexpress, C188-9 CAS:432001-19-9 Purity:>98%
SDP
Catalog No. 501969073
Encompass_Preferred
Medchemexpress, HY-112288 5mg C188-9 CAS:432001-19-9 C188-9 is a Stat3 inhibitor, with a Kd of 4.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9073 Supplier Medchemexpress LLC Supplier No. HY1122885MG
Medchemexpress, HY-112288 5mg C188-9 CAS:432001-19-9 C188-9 is a Stat3 inhibitor, with a Kd of 4.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.