Learn More
Selleck Chemical LLC C188-9 5mg 432001-19-9
SDP
Supplier:  Selleck Chemical LLC S86055MG
Encompass_Preferred
C188-9 is a potent inhibitor of STAT3 that binds to STAT3 with high affinity (KD=4.7¿0.4 nM). C188-9 is well tolerated in mice, shows good oral bioavailability, and is concentrated in tumors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7981