Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C18H14Cl2F2N4O2S | 2505339-54-6 | MFCD34565708 | 99.8% | 459.30 | C18H14Cl2F2N4O2S | 5 MG
SDP

Catalog No. 5000280983
Encompass_Preferred

This selective small-molecule inhibitor targets fibroblast activation protein (FAP) and is intended for research use. It exhibits potent FAP inhibition in vitro (IC50 = 2 nM), shows dose-dependent activity in animal studies, and is supplied as a high-purity solid with defined storage recommendations.

  • Selective fibroblast activation protein inhibitor with IC50 of 2 nM.
  • Also inhibits prolyl oligopeptidase with lower potency (IC50 ≈ 49 μM).
  • High purity, ≥99.82% by HPLC.
  • Molecular weight 459.30 g/mol and formula C18H14Cl2F2N4O2S.
  • Appearance: white to off-white solid.
  • Storage: keep sealed, away from moisture and light; in solvent -80°C (6 months), -20°C (1 month).
  • Supplied as a small quantity suitable for in vitro and in vivo research.
  • For research use only; not for human or diagnostic use.

Catalog No. 50-002-80983 Supplier Medchemexpress LLC Supplier No. HYW4040025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This selective small-molecule inhibitor targets fibroblast activation protein (FAP) and is intended for research use. It exhibits potent FAP inhibition in vitro (IC50 = 2 nM), shows dose-dependent activity in animal studies, and is supplied as a high-purity solid with defined storage recommendations.

  • Selective fibroblast activation protein inhibitor with IC50 of 2 nM.
  • Also inhibits prolyl oligopeptidase with lower potency (IC50 ≈ 49 μM).
  • High purity, ≥99.82% by HPLC.
  • Molecular weight 459.30 g/mol and formula C18H14Cl2F2N4O2S.
  • Appearance: white to off-white solid.
  • Storage: keep sealed, away from moisture and light; in solvent -80°C (6 months), -20°C (1 month).
  • Supplied as a small quantity suitable for in vitro and in vivo research.
  • For research use only; not for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.